Learn More
Description
- Tool for cleaning walls and tanks
- Ideal for scrubbing stubborn dirt and cleaning underneath equipment
- The clip mechanism is stainless steel, which prevents the pad from falling off once heavy with water
- Color-coded
Specifications
Specifications
| Sterility | Non-sterile |
| Color | White |
| Diameter (English) | 3.8 in. |
| Dimensions (L x W x H) | 9.1 x 3.8 x 3 in. |
| Material | Polypropylene, Stainless Steel, Nylon |
| Quantity | 1 Ea. |
| Size | Medium |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.