Learn More
Description
- With drip-free spout
- Hand-grip raised from base and stainless-steel handle
- Calibrated for variety of measuring units
- Flat side prevents spillage
- Stores using coordinating wall bracket
Specifications
Specifications
| Product Type | Transporting Pail |
| Material | Polypropylene |
| Capacity (English) | 3 gal. |
| Color | Yellow |
| For Use With (Application) | transporting cleaning chemicals, hot or cold ingredients |
| Temperature (Metric) | 121°C |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.