Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Vikan™ Liquid Transporting Pail

Catalog No. 19830035 Shop All Remco Products
Change view
Click to view available options
Color:
Black
Red
White
Yellow
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
19-830-035 White
19-830-037 Red
19-830-034 Yellow
19-830-030 Black
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 19-830-035 Supplier Vikan™ Supplier No. 56865
Add to Cart
Add to Cart

For transporting cleaning chemicals and hot or cold ingredients.

  • With drip-free spout
  • Hand-grip raised from base and stainless-steel handle
  • Calibrated for variety of measuring units
  • Flat side prevents spillage
  • Stores using coordinating wall bracket

Specifications

Product Type Transporting Pail
Material Polypropylene
Capacity (English) 3 gal.
Color White
For Use With (Application) transporting cleaning chemicals, hot or cold ingredients
Temperature (Metric) 121°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.