Learn More
Vikan™ Long Handle Scrubbing Brush
Catalog No. 17227282 Shop All Remco Products
Change view
Click to view available options
Color:
Black
Blue
Green
Orange
Pink
Purple
Red
White
Yellow
9 product options available for selection
Product selection table with 9 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
17-227-282 Red
17-227-284 Orange
17-227-283 Yellow
17-227-280 Green
17-227-281 Blue
17-227-285 Purple
17-227-279 Pink
19-830-224 White
19-830-247 Black
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 9 options available.
9 options
Catalog No. 17-227-282 Supplier Vikan™ Supplier No. 41864
Add to Cart
Add to Cart
Vikan™ Long Handle Scrubbing Brush is a long handled scrubbing brush with stiff bristles.
  • Allows you to reach into parts of equipment that are hard to access
  • Angled bristles are stiff, helping to scrub at a distance
  • Color-coded
Specifications
Product Type Scrub Brush
Color-coded Yes
Handle Material Polypropylene
Sterility Non-sterile
Certifications/Compliance FDA Compliant
Color Red
Length (English) 15.75 in.
Quantity 1 Ea.
Size Medium