Description
- Smooth handle
- Ideal for cleaning stubborn food soils in narrow areas such as around conveyor belts, rubber seals of fridges, baker's racks, and other tight spaces
- Can also used to clean fish
- Color-coded
Specifications
Specifications
| Product Type | Utility Brush |
| Color-coded | Yes |
| Handle Material | Polypropylene |
| Sterility | Non-sterile |
| Certifications/Compliance | FDA Compliant |
| Color | Green |
| Length (English) | 11.8 in. |
| Quantity | 1 Ea. |
| Size | Narrow |