Learn More
Vikan™ Narrow Utility Brush
Catalog No. 17227307 Shop All Remco Products
Change view
Click to view available options
Color:
Blue
Green
Red
White
Yellow
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
17-227-307 White
17-227-306 Red
17-227-308 Yellow
17-227-304 Green
17-227-305 Blue
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 17-227-307 Supplier Vikan™ Supplier No. 41955
Only null left
Add to Cart
Add to Cart
Vikan™ Narrow Utility Brush is a cleaning brush with a smooth, egronomically designed handle.
  • Smooth handle
  • Ideal for cleaning stubborn food soils in narrow areas such as around conveyor belts, rubber seals of fridges, baker's racks, and other tight spaces
  • Can also used to clean fish
  • Color-coded
Specifications
Product Type Utility Brush
Bristle Material Polyester
Bristle Type Extra Stiff
Color-coded Yes
Handle Material Polypropylene
Sterility Non-sterile
Certifications/Compliance FDA Compliant
Color White
Length (English) 11.8 in.
Quantity 1 Ea.
Size Narrow