Learn More
Vikan™ Short Handle Utility Brush
Catalog No. 17227288 Shop All Remco Products
Change view
Click to view available options
Color:
Green
Red
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
17-227-288 Red
17-227-286 Green
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 17-227-288 Supplier Vikan™ Supplier No. 41904
Vikan™ Short Handle Utility Brush is a short-handled washing brush with medium bristles.
  • Comfortable grip
  • Allows you to work for long periods of time without hand fatigue
  • Medium strength bristles are great for washing all types of equipment, tables, and walls
  • Brush can be used holding both the handle and the brush head for better control
  • Color-coded
Specifications
Product Type Utility Brush
Color-coded Yes
Handle Material Polypropylene
Sterility Non-sterile
Certifications/Compliance FDA Compliant
Color Red
Length (English) 9.8 in.
Quantity 1 Ea.
Size Short