Learn More
Vikan™ Soft UST Detail Brush
Catalog No. 17227487 Shop All Remco Products
Change view
Click to view available options
Color:
Blue
Green
Orange
Pink
Purple
Red
White
Yellow
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
17-227-487 Purple
17-227-483 Red
17-227-486 Orange
17-227-485 Yellow
17-227-481 Green
17-227-482 Blue
17-227-480 Pink
17-227-484 White
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
Catalog No. 17-227-487 Supplier Vikan™ Supplier No. 5551308
Add to Cart
Add to Cart
Vikan™ Soft UST Detail Brush is designed for detail cleaning in dry environments where soft bristles are useful and hygiene is critical.
  • Uses Ultra Safe Technology to reinvent detail cleaning
  • Extends the benefits of UST to every nook and cranny in your plant
  • Soft bristles are ideal for food contact approval, pastry or glazing applications, amd other food-handling applications
  • Color-coded
Specifications
Product Type Cleaning Brush
Color-coded Yes
Handle Material Polypropylene
Sterility Non-sterile
Certifications/Compliance FDA Compliant
Color Purple
Length (English) 8.07 in.
Quantity 1 Ea.
Size Small