Learn More
Vikan™ Spray Gun
Catalog No. 17227981 Shop All Remco Products
Change view
Click to view available options
Color Code:
Black
Blue
Green
Red
White
Yellow
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color Code
17-227-981 Yellow
17-227-979 Red
17-227-977 Green
17-227-978 Blue
17-227-980 White
17-227-982 Black
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 17-227-981 Supplier Vikan™ Supplier No. 93246
Add to Cart
Add to Cart
Spray gun allows for a gentle flow or a high-pressured stream of water from an attached hose to clean different surfaces.
  • Suitable for cleaning floors, work surfaces, walls, and machinery
  • By applying a variable amount of tension on the rubber insulated trigger, the user can control the pressure
  • The tool is suitable for many cleaning applications, including rinsing off work surfaces and walls after chemical foaming
  • The water jet can also be adjusted using the screw at the end of gun
  • An arrow on the water gun indicates the spray direction of the water jet
  • Included adapter fits to our Automatic Shut Off Coupling (0703US) or the Hose Clamp (0701) can be used to attach it directly to a hose
Specifications
Product Type Spray Gun
Color Code Yellow
Color-coded Yes
Material Rubber, Brass, Stainless Steel
Size Medium
Dimensions (L x W) 5.7 x 7.1 in.
No. per Pack 1 Ea.
Weight (English) 1.76 lb.