Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Vikan™ Standard Broom Handle

Catalog No. 19830017 Shop All Remco Products
Change view
Click to view available options
Color Code:
Black
Red
White
Yellow
Length (English):
51 in.
60 in.
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color Code Length (English)
19-830-017 White 60 in.
19-830-015 Black 60 in.
19-830-016 Yellow 60 in.
19-830-018 Red 60 in.
19-830-019 Yellow 51 in.
19-830-020 Black 51 in.
19-830-021 Red 51 in.
19-830-022 White 51 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
Catalog No. 19-830-017 Supplier Vikan™ Supplier No. 29375
Add to Cart
Add to Cart

The handle with attached item reaches to the chin of the user to prevent occupational back injuries.

  • Made from aluminum and polypropylene
  • Used with brooms, squeegees and scrapers

Specifications

Color Code White
Product Type Standard Broom Handle
Length (English) 60 in.
Autoclavable Autoclavable
For Use With (Equipment) Brooms, Squeegees and Scrapers
Description Hygienic (Euro) Thread
Color-coded Yes
Material Aluminum, Polypropylene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.