Learn More
Description
- Made from aluminum and polypropylene
- Used with brooms, squeegees and scrapers
Specifications
Specifications
| Color Code | Black |
| Product Type | Standard Broom Handle |
| Length (English) | 51 in. |
| Autoclavable | Autoclavable |
| For Use With (Equipment) | Brooms, Squeegees and Scrapers |
| Description | Hygienic (Euro) Thread |
| Color-coded | Yes |
| Material | Aluminum, Polypropylene |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.