Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Vikan™ Stiff 12 in. Deck/Wall Scrub

Catalog No. 17227807 Shop All Remco Products
Change view
Click to view available options
Color:
Black
Blue
Green
Orange
Pink
Purple
Red
White
Yellow
9 product options available for selection
Product selection table with 9 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
17-227-807 Black
17-227-802 Red
17-227-805 Orange
17-227-804 Yellow
17-227-800 Green
17-227-801 Blue
17-227-806 Purple
17-227-799 Pink
17-227-803 White
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 9 options available.
9 options
Catalog No. 17-227-807 Supplier Vikan™ Supplier No. 70609
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Vikan™ Stiff 12 in. Deck/Wall Scrub can be used in conjunction with water and cleaning agents.

  • Scrubbing broom with firm bristles
  • Bristle length is ideal for scrubbing all types of floors; epoxy, tiles, cement, etc
  • Reaches well into holes and grouting to remove hidden dirt
  • Color-coded

Specifications

Product Type Scrub Brush
Color-coded Yes
Handle Material Polypropylene
Sterility Non-sterile
Certifications/Compliance FDA Compliant
Color Black
Description Thread type Euro Hygienic
Length (English) 11.8 in.
Quantity 1 Ea.
Size Large
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.