Learn More
Vikan™ Stiff Tank Brush
Catalog No. 17227763 Shop All Remco Products
Change view
Click to view available options
Color:
Black
Blue
Green
Orange
Purple
Red
White
Yellow
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
17-227-763 Black
17-227-758 Red
17-227-761 Orange
17-227-760 Yellow
17-227-756 Green
17-227-757 Blue
17-227-762 Purple
17-227-759 White
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
Catalog No. 17-227-763 Supplier Vikan™ Supplier No. 70379
Add to Cart
Add to Cart
Vikan™ Stiff Tank Brush is ideal for scrubbing inside large industrial tanks and other large container-like equipment.
  • Oval-shaped brush
  • Stiff bristles
  • Effective for scrubbing inside large industrial tanks and other large container-like equipment
  • Rounded edges work well to clean down the sides of equipment
  • Fits well into large channel drains
  • Color-coded
Specifications
Product Type Cleaning Brush
Color-coded Yes
Handle Material Polypropylene
Sterility Non-sterile
Certifications/Compliance FDA Compliant
Color Black
Description Thread type Euro Hygienic
Length (English) 8.3 in.
Quantity 1 Ea.
Size Large