Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Vikan™ Ultra Hygiene Handles

Catalog No. 19830054 Shop All Remco Products
missing translation for 'changeView'
missing translation for 'orderingAttributeHoverText'
Color Code:
Blue
Green
Red
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color Code
19-830-054 Blue
19-830-053 Green
19-830-055 Red
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 missing translation for 'options'
Catalog No. 19-830-054 missing translation for 'mfr' Vikan™ missing translation for 'supplierNo' 29603
Add to Cart
Add to Cart

Used with brooms, squeegees and scrapers.

  • Made from chemically resistant polypropylene
  • Can be safely used in a variety of environments
  • Hygienic design minimizes the risk of cross-contamination making it easy to clean and maintain
  • Autoclavable to 250°F/121°C

Specifications

Color Code Blue
Product Type Ultra Hygiene Handle
Length (English) 51 in.
Width (English) 1.25 in.
Color-coded Yes
Material Polypropylene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.