Learn More
Medchemexpress LLC Vilazodone-d8 | 1794789-93-7 | MFCD28900708 | 99.0% | 449.57 g/mol | C26H19D8N5O2 | 5 MG

Supplier: Medchemexpress LLC HY14261S5MG
Vilazodone-d8 is the deuterium-labeled analogue of vilazodone supplied for use as a reference standard in analytical, pharmacokinetic, and tracer studies. It preserves the pharmacological profile of vilazodone (a serotonin selective reuptake inhibitor and 5-HT1A receptor partial agonist) and is intended primarily as an internal standard for LC-MS quantitation and related research applications.
- Deuterium-labeled analogue (d8) suitable as an internal standard.
- High purity, 99.0%.
- Molecular formula C26H19D8N5O2; molecular weight 449.57 g/mol.
- Available in small research pack sizes appropriate for analytical workflows.
- Stable when stored as powder at -20°C or 4°C for specified durations.
- Solubility in water ≥ 0.32 mg/mL (0.71 mM), compatible with common LC solvents.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.