Learn More
Vileda Professional™ CE Flat Sponge Mop Refill with Polyester Lamination, Sterile
Shop All Vileda Professional ProductsDescription
- Features polyester lamination for tear-free application of disinfectants, cleaning and spill absorption
- Validated to 10-6 SAL per ISO 11137
- Individually double bagged with Lot # and Indicator Dot on each package
- Certificate of Sterility available with each shipment, upon request
Specifications
Specifications
| Autoclavable | Autoclavable |