Learn More
Description
The MA511883A555 immunogen is purified vimentin from pig eye lens.
Specifications
Specifications
| Antigen | Vimentin |
| Applications | Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry |
| Classification | Monoclonal |
| Clone | V9 |
| Concentration | 1 mg/mL |
| Conjugate | Alexa Fluor 555 |
| Formulation | PBS with 1mg/mL BSA and 0.05% sodium azide; pH 7.4 |
| Gene | Vim |
| Gene Accession No. | P02543, P08670, P09654, P20152, P31000, P48616, P48670 |
| Gene Alias | cb28; class-III intermediate microfilament; CTRCT30; epididymis luminal protein 113; FLJ36605; Hel113; intermediate filament; intermediate filament protein; RP11-124N14.1; similar to vimentin; VIM; VIME; Vimentin; vimentin C-terminus |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
