Learn More
Description
This antibody reacts with human vimentin, a 57 kDa intermediate filament intracellular protein expressed on a wide variety of mesenchymal and mesodermal cell types. This antibody will not cross-react with porcine or mouse.
Specifications
Specifications
| Antigen | Vimentin |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Clone | VI-RE/1 |
| Concentration | 0.1 mg/mL |
| Conjugate | PE |
| Formulation | PBS with 0.2% BSA and 15mM sodium azide; pH 7.4 |
| Gene | Vim |
| Gene Accession No. | P08670 |
| Gene Alias | cb28; class-III intermediate microfilament; CTRCT30; epididymis luminal protein 113; FLJ36605; Hel113; intermediate filament; intermediate filament protein; RP11-124N14.1; similar to vimentin; VIM; VIME; Vimentin; vimentin C-terminus |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
