Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Vinblastine sulfate, 100.2%, MP Biomedicals™

Catalog No. ICN19028701 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
1 mg
5 mg
10 mg
25 mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN19028701 1 mg
ICN19028705 5 mg
ICN19028710 10 mg
ICN19028725 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. ICN19028701 Supplier MP Biomedicals, Inc Supplier No. 0219028701
Add to Cart
Add to Cart

Chemical Identifiers

CAS 143-67-9
Molecular Formula C46H60N4O13S
Molecular Weight (g/mol) 909.06
MDL Number MFCD08706468,MFCD00082457,MFCD08706468
InChI Key KDQAABAKXDWYSZ-JKDPCDLQSA-N
Synonym vinblastine sulfate
PubChem CID 131709225
IUPAC Name methyl (1R,9R,10S,11R,12R,19R)-11-(acetyloxy)-12-ethyl-4-[(13S,15S,17S)-17-ethyl-17-hydroxy-13-(methoxycarbonyl)-1,11-diazatetracyclo[13.3.1.0⁴,¹².0⁵,¹⁰]nonadeca-4(12),5,7,9-tetraen-13-yl]-10-hydroxy-5-methoxy-8-methyl-8,16-diazapentacyclo[10.6.1.0¹,⁹.0²,⁷.0¹⁶,¹⁹]nonadeca-2(7),3,5,13-tetraene-10-carboxylate; sulfuric acid
SMILES OS(O)(=O)=O.CC[C@]1(O)C[C@H]2CN(C1)CCC1=C(NC3=CC=CC=C13)[C@@](C2)(C(=O)OC)C1=CC2=C(C=C1OC)N(C)[C@@H]1[C@]22CCN3CC=C[C@](CC)([C@@H]23)[C@@H](OC(C)=O)[C@]1(O)C(=O)OC

Specifications

Color White to Yellow
Quantity 1 mg
Formula Weight 909.1
Physical Form Powder
Chemical Name or Material Vinblastine sulfate
EINECSNumber 205-606-0
Recommended Storage Store at +4°C
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.