CAS RN 143-67-9
Thermo Scientific Chemicals Vinblastine Sulfate, 98%
CAS: 143-67-9 Molecular Formula: C46H60N4O13S Synonym: Vincaleukblastine sulfate
Vinblastine sulfate, 100.2%, MP Biomedicals™
CAS: 143-67-9 Molecular Formula: C46H60N4O13S Molecular Weight (g/mol): 909.06 MDL Number: MFCD08706468,MFCD00082457,MFCD08706468 InChI Key: KDQAABAKXDWYSZ-JKDPCDLQSA-N Synonym: vinblastine sulfate PubChem CID: 131709225 IUPAC Name: methyl (1R,9R,10S,11R,12R,19R)-11-(acetyloxy)-12-ethyl-4-[(13S,15S,17S)-17-ethyl-17-hydroxy-13-(methoxycarbonyl)-1,11-diazatetracyclo[13.3.1.0⁴,¹².0⁵,¹⁰]nonadeca-4(12),5,7,9-tetraen-13-yl]-10-hydroxy-5-methoxy-8-methyl-8,16-diazapentacyclo[10.6.1.0¹,⁹.0²,⁷.0¹⁶,¹⁹]nonadeca-2(7),3,5,13-tetraene-10-carboxylate; sulfuric acid SMILES: OS(O)(=O)=O.CC[C@]1(O)C[C@H]2CN(C1)CCC1=C(NC3=CC=CC=C13)[C@@](C2)(C(=O)OC)C1=CC2=C(C=C1OC)N(C)[C@@H]1[C@]22CCN3CC=C[C@](CC)([C@@H]23)[C@@H](OC(C)=O)[C@]1(O)C(=O)OC
Thermo Scientific Chemicals Vinblastine sulfate, 96.0-102.0%
CAS: 143-67-9 Molecular Formula: C46H60N4O13S Molecular Weight (g/mol): 909.06 MDL Number: MFCD08706468,MFCD00082457,MFCD08706468 InChI Key: KDQAABAKXDWYSZ-JKDPCDLQSA-N Synonym: vinblastine sulfate PubChem CID: 131709225 IUPAC Name: methyl (1R,9R,10S,11R,12R,19R)-11-(acetyloxy)-12-ethyl-4-[(13S,15S,17S)-17-ethyl-17-hydroxy-13-(methoxycarbonyl)-1,11-diazatetracyclo[13.3.1.0⁴,¹².0⁵,¹⁰]nonadeca-4(12),5,7,9-tetraen-13-yl]-10-hydroxy-5-methoxy-8-methyl-8,16-diazapentacyclo[10.6.1.0¹,⁹.0²,⁷.0¹⁶,¹⁹]nonadeca-2(7),3,5,13-tetraene-10-carboxylate; sulfuric acid SMILES: OS(O)(=O)=O.CC[C@]1(O)C[C@H]2CN(C1)CCC1=C(NC3=CC=CC=C13)[C@@](C2)(C(=O)OC)C1=CC2=C(C=C1OC)N(C)[C@@H]1[C@]22CCN3CC=C[C@](CC)([C@@H]23)[C@@H](OC(C)=O)[C@]1(O)C(=O)OC
MilliporeSigma™ Calbiochem™ Vinblastine Sulfate
Antitumor alkaloid drug.
R&D Systems™ Vinblastine sulfate
CAS: 143-67-9 Molecular Formula: C46H60N4O13S