Learn More
Novus Biologicals™ Vinblastine sulfate

Description
Biological Activity:Potent anticancer agent that blocks DNA synthesis. Induces apoptosis via p53-dependent and -independent mechanisms. Inhibits X-linked inhibitor of apoptosis protein (XIAP) expression and activates caspase-3. In certain glioma cell lines, sensitizes cells to TNF-a-induced apoptosis.
Storage: Store at 4°C.
Note: Inhibitors are guaranteed for 1 year from date of receipt.
Specifications
Specifications
| Formulation | C46H58N4O9.H2SO4 |
| For Use With (Application) | Inhibits apoptosis |
| Quantity | 50 mg |
| Product Type | Inhibitors |
| Molecular Weight (g/mol) | 909.06g/mol |
| Solubility | Soluble to 100mM in DMSO, 100mM water |
| Content And Storage | 4°C |
| Target | Vinblastine sulfate |
| Color | White |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.