Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Vincristine sulfate, 99.4%, MP Biomedicals™

Catalog No. ICN19068701 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
1 mg
5 mg
25 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN19068701 1 mg
ICN19068705 5 mg
ICN19068725 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. ICN19068701 Supplier MP Biomedicals, Inc Supplier No. 0219068701
Add to Cart
Add to Cart

Chemical Identifiers

CAS 2068-78-2
Molecular Formula C46H58N4O14S
Molecular Weight (g/mol) 923.044
InChI Key AQTQHPDCURKLKT-DJOZZVTQSA-N
Synonym vincristine sulfate, methyl 9r,10s,11r,12r,19r-11-acetyloxy-12-ethyl-4-13s,15r,17s-17-ethyl-17-hydroxy-13-methoxycarbonyl-1,11-diazatetracyclo 13.3.1.0?, 1 2 .0?, 1 ? nonadeca-4 12 ,5,7,9-tetraen-13-yl-8-formyl-10-hydroxy-5-methoxy-8,16-diazapentacyclo 10.6.1.0 1 ,?.0 2 ,?.0
PubChem CID 126963557
SMILES CCC1(CC2CC(C3=C(CCN(C2)C1)C4=CC=CC=C4N3)(C5=C(C=C6C(=C5)C78CCN9C7C(C=CC9)(C(C(C8N6C=O)(C(=O)OC)O)OC(=O)C)CC)OC)C(=O)OC)O.OS(=O)(=O)O

Specifications

Color White
Assay Percent Range 0.994
Quantity 1 mg
Formula Weight 923
Physical Form Powder
Chemical Name or Material Vincristine sulfate
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.