Learn More
Selleck Chemical LLC Estramustine phosphate sodium S2441-25mg
Supplier: Selleck Chemical LLC S244125MG
Estramustine phosphate sodium (EMP Emcyt Estracyt) is an antitumour drug with a unique dual mode of action Estrone and estradiol products of the metabolism of estramustine phosphate sodium have shown antigonadotrophic activity resulting in reduced testosterone levels Estramustine(the cytotoxic metabolite produced by dephosphorylation of estramustine phosphate sodium) is an antiprostatic tumor drug with antimicrotubule effects
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.