Learn More
Selleck Chemical LLC Vinorelbine Tartrate 10mg 125317-39-7
SDP
Supplier:  Selleck Chemical LLC S426910MG
Encompass_Preferred
Vinorelbine Tartrate is a semi-synthetic vinca alkaloid, and inhibits mitosis through interaction with tubulin. Vinorelbine Tartrate exhibits anti-tumor activities via inducing the mitotic apoptosis, autophagy and inflammation. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-136-3823
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.