Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Vinyl benzoate, 95%

Catalog No. AAB2180522
Change view
Click to view available options
Quantity:
100 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2180522 100 g
AAB2180536 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2180522 Supplier Thermo Scientific Chemicals Supplier No. B2180522
Add to Cart

CAS: 769-78-8 | C9H8O2 | 148.161 g/mol

It is used as chemical reagent and pharmaceutical intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used as chemical reagent and pharmaceutical intermediates.

Solubility
Soluble in methanol

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 769-78-8
Molecular Formula C9H8O2
Molecular Weight (g/mol) 148.161
MDL Number MFCD00048141
InChI Key KOZCZZVUFDCZGG-UHFFFAOYSA-N
Synonym vinyl benzoate, benzoic acid vinyl ester, benzoic acid, ethenyl ester, benzoic acid, vinyl ester, unii-f1e7c1ggku, vinylester kyseliny benzoove, vinylester kyseliny benzoove czech, f1e7c1ggku, benzoic acid, ethenyl ester 9ci, benzoic acid vinyl
PubChem CID 13037
ChEBI CHEBI:84279
IUPAC Name ethenyl benzoate
SMILES C=COC(=O)C1=CC=CC=C1

Specifications

Density 1.07
Boiling Point 73°C to 74°C (8 mmHg)
Flash Point 82°C (180°F)
Refractive Index 1.529
Quantity 100 g
Beilstein 2041125
Sensitivity Moisture sensitive
Solubility Information Soluble in methanol
Formula Weight 148.16
Percent Purity 95%
Chemical Name or Material Vinyl benzoate
Show More Show Less
Hazard Category H227-H317
Hazard Statement GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.
Precautionary Statement P210-P261-P272-P280-P302+P352-P333+P313-P363-P370+P378q-P501c
EINECSNumber 212-214-3
RTECSNumber DI1050000
TSCA No
Recommended Storage Ambient temperatures; Store under Argon

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.