Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Vinyl Bromide (ca. 12% in Tetrahydrofuran, ca. 1.0mol/L), TCI America™
SDP

Catalog No. V0127100ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
100 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
V0127100ML 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. V0127100ML Supplier TCI America Supplier No. V0127100ML
Add to Cart
Add to Cart

Chemical Identifiers

CAS 593-60-2
Molecular Formula C2H3Br
Molecular Weight (g/mol) 106.95
MDL Number MFCD00000183
InChI Key INLLPKCGLOXCIV-UHFFFAOYSA-N
Synonym vinyl bromide, bromoethylene, ethene, bromo, monobromoethylene, vinylbromid, bromure de vinyle, vinylbromide, polyvinyl bromide, ethylene, bromo, polyvinylbromide
PubChem CID 11641
ChEBI CHEBI:51311
IUPAC Name bromoethene
SMILES BrC=C

Specifications

Color Colorless
Quantity 100 mL
UN Number 1993
Formula Weight 106.95
Physical Form Liquid
Chemical Name or Material Vinyl Bromide (ca. 12% in Tetrahydrofuran, ca. 1.0mol/L)
RTECSNumber KU8400000
TSCA Yes
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.