Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Vinyl Butyrate (stabilized with MEHQ) 98.0+%, TCI America™
SDP

Catalog No. B0765100ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 mL
100 mL
500 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B0765100ML 100 mL
B076525ML 25 mL
B0765500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. B0765100ML Supplier TCI America Supplier No. B0765100ML
Add to Cart
Add to Cart

Chemical Identifiers

CAS 123-20-6
Molecular Formula C6H10O2
Molecular Weight (g/mol) 114.144
MDL Number MFCD00059410
InChI Key MEGHWIAOTJPCHQ-UHFFFAOYSA-N
Synonym Butyric Acid Vinyl Ester
PubChem CID 31247
IUPAC Name ethenyl butanoate
SMILES CCCC(=O)OC=C

Specifications

Color Colorless
Boiling Point 116°C
Quantity 100 mL
UN Number 2838
Formula Weight 114.14
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Vinyl Butyrate (stabilized with MEHQ)
EINECSNumber (2)-3697
RTECSNumber ET7000000
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.