Learn More
Description
- Rubber adhesive sticks fast and stays fast on many surfaces, even irregular surfaces
- Embossed vinyl backing seals out dust, dirt, and moisture, and resists abrasion and many chemicals
- Ideal for a variety of purposes including temporary repairs, identification, contrast, decoration, securing wiring, bundling, sealing, holding, protecting, and other jobs including applications requiring moisture resistance
- Peels off cleanly after 30 minutes on painted steel as hot as 200°F (93°C)
- Easy to tear and conforms to irregular surfaces
- Available in a variety of colors for coding, process tagging, highlighting trip hazards, and enhancing décor
Specifications
Specifications
| Color | Green |
| Product Type | Vinyl Duct Tape |
| For Use With (Application) | Communications, Design and Construction, Manufacturing, Mining, Oil and Gas, Transportation |
| Model | 3903 |
| Quantity | 24/Cs. |
| Thickness (English) | 6.5 mil |
| Dimensions (L x W) | 50 yd. x 2 in. |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.