Learn More
Description
- Vinyl laurate acts as an acyl donor during the transeterification of L-α-glycerophosphorylcholine (gpc) in the presence of candida antarctica lipase b as catalyst to form lysophosphatidylcholine (lpc).vinyl laurate can be used to synthesize the following:
- Sucrose laurate esters from sucrose in organic solvents
- Poly(vinyl laurate) (pvl)latexes
- Porous conducting soft polymer foams which can be used as gas sensors
Specifications
Specifications
| CAS | 2146-71-6 |
| Density | 0.871 g/mL (at 20°C (literature)) |
| Boiling Point | 254°C (1013 hPa (lit.)) |
| Molecular Formula | C14H26O2 |
| Refractive Index | n20/D 1.441 |
| Linear Formula | CH3(CH2)10 COOCH=CH2 |
| MDL Number | MFCD00048421 |
| Quantity | 1 L |
| Molecular Weight (g/mol) | 226.36 |
| Percent Purity | ≥99.0% (GC) |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.