Learn More
Sigma Organic Chemistry Vinyl propionate | 500ML | 105-38-4 | MFCD00027004 | 0.98
Supplier: Sigma Organic Chemistry 401714500ML

Vinyl propionate, 500ML
About This Item:
Synonym(s): Vinyl propanoate
Linear Formula: CH3CH2COOCH=CH2
Molecular Weight: 100.12
CAS Number: 105-38-4
MDL Number: MFCD00027004
UNSPSC Code: 12162002
Purity: 0.98
Boiling Point: 94-95 C (lit.)
Melting Point: -80 C (lit.)
Density: 0.919 g/mL at 25 C (lit.)
Refractive Index: n20/D 1.403 (lit.)
Storage: 2-8C
EINECS Number: 203-293-5
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.