Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Brady™ Vinyl Sign: WARNING - PESTICIDE SPRAYING, B946, 20 x 14 in., Black/Red/White

Catalog No. 19106763 Shop All Brady Corporation Products
Change view
Click to view available options
Dimensions (L x W):
14 x 20 in.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Dimensions (L x W)
19-106-763 14 x 20 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19-106-763 Supplier Brady™ Supplier No. 17530

Improve workplace safety with proper signage and markings. This Brady™ vinyl sign is a durable, clear and effective tool to raise awareness to workplace hazards.

  • Vinyl material is abrasion-resistant and chemical-resistant
  • Safety signs are important facility visuals that are vital to communicating warnings and other safety information
  • Self sticking vinyl will adhere to difficult low surface energy items, including: PVC piping, blow-molded equipment cases, ABS plastics, powder-coated surfaces and equipment made from recycled plastics
  • Excels at sticking to highly textured surfaces such as painted cinder block walls, finished uneven wood, textured plastics, rough-pipes and paper-jacketed pipes.
  • Bold and bright, providing high visibility to critical messages
  • For indoor and outdoor use

Specifications

Material Vinyl
Shape Rectangular
Dimensions (L x W) 14 x 20 in.
Displayed Text WARNING - PESTICIDE SPRAYING
Color Red on White

Adhere to
Safety Regulations and Best Practices

Take advantage of a range of Safety Services from our trusted suppliers to help ensure the safety and compliance of your people, processes, products, and facilities. Services include assessments, training, equipment maintenance, safety programs, and more.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.