Learn More
Description
- Available in a wide variety of vivid colors for diverse applications
- Unique stretch properties enable tape to remain stretched to conform to uneven surfaces without lifting and retracting
- Designed to resist wear, scrapes and moisture
- One-piece, clean removal from many surfaces which helps reduce clean-up and labor costs
- Rubber adhesive provides excellent holding strength on many surfaces
- Good abrasion and solvent resistance for long service life
Specifications
Specifications
| Packaging Quantity | 144 Rolls/Cs. |
| Dimensions (L x W) | 36 yd. x 1/4 in. |
| Thickness | 5.2 mil |
| Color | Orange |
| For Use With (Application) | Color Coding Tools and Equipment, Electroplating and Anodizing Processes, Fine Line Masking, Floor |
| Adhesive Type | Rubber |
| Material | Vinyl |
| Product Type | Flagging Tape |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.