Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Vinylene Carbonate (stabilized with BHT) 98.0+%, TCI America™
SDP

Catalog No. V001525G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
V001525G 25 g
V00155G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. V001525G Supplier TCI America Supplier No. V001525G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 872-36-6
Molecular Formula C3H2O3
Molecular Weight (g/mol) 86.05
MDL Number MFCD00005380
InChI Key VAYTZRYEBVHVLE-UHFFFAOYSA-N
Synonym vinylene carbonate, vinyl carbonate, carbonic acid, cyclic vinylene ester, wln: t5ovoj, vinylenecarbonate, vinyleny carbonate, carbonic acid vinylene, 1,3-dioxo-2-one, vinylene carbonate vc, acmc-209qj3
PubChem CID 13385
IUPAC Name 2H-1,3-dioxol-2-one
SMILES O=C1OC=CO1

Specifications

Color White-Yellow
Boiling Point 60°C
Quantity 25 g
Formula Weight 86.05
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Lumps
Chemical Name or Material Vinylene Carbonate (stabilized with BHT)
EINECSNumber (5)-6636&(5)-6744
RTECSNumber FG3325000
TSCA Yes
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.