Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Vinylmagnesium bromide, 1M solution in 2-MeTHF, AcroSeal™

Catalog No. AC445971000
Change view
Click to view available options
Quantity:
100 mL
800 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC445971000 100 mL
AC445978000 800 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC445971000 Supplier Thermo Scientific Chemicals Supplier No. 445971000
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

CAS: 1826-67-1 | C2H3BrMg | 131.26 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1826-67-1 , 96-47-9
Molecular Formula C2H3BrMg
Molecular Weight (g/mol) 131.26
MDL Number MFCD00000042
InChI Key XHHHAXOHMKAOSL-UHFFFAOYSA-M
Synonym vinylmagnesium bromide, bromo ethenyl magnesium, vinyl magnesium bromide, magnesium, bromoethenyl, vinylmagnesium bromide solution, vinylmagnesium bromide solution, 1.0 m in thf, bromovinylmagnesium, bromo vinyl magnesium, vinyl magnesiumbromide
PubChem CID 74584
IUPAC Name magnesium;ethene;bromide
SMILES Br[Mg]C=C

Specifications

Density 0.9650g/mL
Flash Point −11°C
Packaging AcroSeal™ Glass bottle
Linear Formula CH2CHMgBr
Quantity 100 mL
Specific Gravity 0.965
Solubility Information Solubility in water: reacts
Formula Weight 131.25
Concentration or Composition (by Analyte or Components) 1M solution in 2-MeTHF
Chemical Name or Material Vinylmagnesium bromide
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Highly flammable liquid and vapor.
In contact with water releases flammable gases which may ignite spontaneously.
Causes severe skin burns and eye damage.
Harmful if swallowed.
Reacts violently with water.<b
Precautionary Statement GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Handle under inert gas.
Protect from moisture
EINECSNumber 217-375-3

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.