Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Vinyltriethylsilane, 97%

Catalog No. p-7045916
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1246103 1 g
AAL1246106 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1246103 Supplier Thermo Scientific Chemicals Supplier No. L1246103
Only null left
Add to Cart

CAS: 1112-54-5 | C8H18Si | 142.32 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1112-54-5
Molecular Formula C8H18Si
Molecular Weight (g/mol) 142.32
MDL Number MFCD00041345
InChI Key HBWGDHDXAMFADB-UHFFFAOYSA-N
Synonym vinyltriethylsilane, triethylvinylsilane, triethyl vinyl silane, vinyl triethyl silane, ethenyl-triethylsilane, triethylsilyl ethylene, ethenyl triethyl silane, pubchem13130, triethyl vinyl silane #
PubChem CID 559195
IUPAC Name ethenyl(triethyl)silane
SMILES CC[Si](CC)(CC)C=C

Specifications

Boiling Point 145°C to 146°C
Quantity 1 g
UN Number UN1993
Beilstein 1743830
Flash Point 25°C (77°F)
Formula Weight 142.32
Percent Purity 97%
Refractive Index 1.434
Density 0.771
Chemical Name or Material Vinyltriethylsilane
Hazard Category H226-H315-H319-H335
Hazard Statement GHS H Statement
H226
Flammable liquid and vapor.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P332+P313-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.