Learn More
Thermo Scientific™ Violet Red Bile Agar
Catalog No. R455282 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
500 g
Catalog No. Quantity
R455282 500 g
1 options
This item is not returnable. View return policy
Catalog No. R455282 Supplier Thermo Scientific™ Supplier No. R455282
Add to Cart
Add to Cart
This item is not returnable. View return policy
The Thermo Scientific™ Remel Violet Red Bile Agar is used for selective isolation and detection of coliform microorganisms in water, foods, and dairy products.
Thermo Scientific™ Remel Violet Red Bile Agar is used for selective isolation and detection of coliform microorganisms in water, foods, milk and other dairy products.
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance Industrial Reference: APHA, BAM, COMPF, IDF.
Description Violet Red Bile Agar
Form Powder
Product Type Agar
Quantity 500 g
Not intended for IVD use.