Learn More
Cytek Biosciences Inc Antibodies violetFluor™ 450 Anti-Mouse CD45.2 (104) ..... Cat. No. 75-0454-U025 ..... Size 25 µg
Supplier:  Cytek Biosciences Inc 750454U025
Encompass_Preferred
Antibodies violetFluor™ 450 Anti-Mouse CD45.2 (104) ..... Cat. No. 75-0454-U025 ..... Size 25 µgThe 104 antibody reacts with mouse CD45.2, also known as Ly5.2, which is a strain-specific allelic form of the CD45 Leukocyte Common Antigen (LCA). Functionally, CD45 is a protein tyrosine phosphatase whose broad cell distribution supports a critical role in many leukocyte functions, including regulation of signal transduction and cell activation associated with the T cell and B cell receptors.The 104 antibody is typically used as a leukocyte marker in Ly5.2 mouse strains C57BL/6, BALB/c, C58, DBA/1, DBA/2, C3H/He, CBA, 129, A and AKR. The antibody has been demonstrated to be specific for CD45.2 and is not cross-reactive with CD45.1-bearing cells. 
Catalog No. 50-210-3643
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.