Learn More
Abcam Viomellein, Antibacterial agent, 1MG
Supplier: Abcam AB1443831MG
MW 560.5 Da, Purity >96%. Gram-positive antibacterial agent. Genotoxic mycotoxin. Dimeric quinone. Interacts with DNA. Shows bactericidal and insecticidal effects and induces mycotoxicosis in vivo. Orally active.
The product is subject to the following: Abcam Restricted Use Statement
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.