Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Scigen VIP-Fixative Leak-Proof Specimen Containers Neutral Buffered 10% Formalin Filled

Catalog No. 23730586 Shop All Scigen Inc Products
Change view
Click to view available options
Capacity (Metric):
120 mL, pre-filled 50%
180 mL, pre-filled 50%
1900 mL, pre-filled 50%
20 mL, pre-filled 50%
40 mL, pre-filled 50%
480 mL, pre-filled 50%
60 mL, pre-filled 50%
7500 mL, pre-filled 50%
90 mL, pre-filled 50%
960 mL, pre-filled 50%
Material:
HDPE
PET
Polypropylene
Color:
Clear
Translucent
White
10 product options available for selection
Product selection table with 10 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (Metric) Material Color
23-730-586 20 mL, pre-filled 50% Polypropylene Translucent
23-730-579 480 mL, pre-filled 50% Polypropylene Translucent
23-730-580 960 mL, pre-filled 50% Polypropylene Translucent
23-730-596 1900 mL, pre-filled 50% PET Clear
23-730-577 7500 mL, pre-filled 50% HDPE White
23-730-587 40 mL, pre-filled 50% Polypropylene Translucent
23-730-588 60 mL, pre-filled 50% Polypropylene Translucent
23-730-589 90 mL, pre-filled 50% Polypropylene Translucent
23-730-590 120 mL, pre-filled 50% Polypropylene Translucent
23-730-591 180 mL, pre-filled 50% Polypropylene Translucent
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 10 options available.
10 options
Catalog No. 23-730-586 Supplier Scigen Supplier No. 1015
Add to Cart
Add to Cart

Safely transport, store and retrieve pathology specimens using Scigen’s Forma-Vial™ system. Containers from 7 mL to 7.5 Liters, filled with 10% neutral buffered formalin, OSHA compliant.

  • Forma-Vial™ specimen containers are uniquely designed to be leak proof and protect against biohazardous or hazardous material release to the environment
  • Highly visible OSHA compliant labeling
  • Sizes range from 7mL dermatology containers to 7.5 Liters operating room organ buckets
  • Container openings range from 13mm to 300mm
  • Filled with 10% neutral buffered formalin to 50% of container’s capacity
  • Formalin waste designed to be treated in bulk using Neutralex or treated individually using Neutralexr tablets (patent pending).
  • Nonsterile containers marked with a lot code and expiration date, 4 year shelf life
  • Manufactured using ISO 13485 and FDA quality systems

Specifications

Product Type Specimen Container Vial
Closure Included Yes
Graduated Yes
Material Polypropylene
Sterility Non-sterile
Capacity (English) 0.7 oz., pre-filled 50%
Capacity (Metric) 20 mL, pre-filled 50%
Closure Color White
Closure Material Polyethylene
Closure Size 37 mm
Closure Type Valve Cap
Color Translucent
Description Specimen Container Pre-filled With Neutral Buffered 10% Formalin
Diameter (Metric) 31 mm
For Use With (Equipment) Histology/cytology specimen containers
Graduations 10 mL
Height (English) 1.7 in.
Height (Metric) 43.5 mm
Includes 50% Filled Neutral Buffered 10% Formalin
Label OSHA Compliant
No. per Case 100/Cs.
No. per Pack 100/Pk.
Product Line Forma-Vial™ Pre-filled Specimen Containers
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.