Learn More
Scigen VIP-Fixative Leak-Proof Specimen Containers Neutral Buffered 10% Formalin Filled
Shop All Scigen Inc ProductsDescription
- Forma-Vial™ specimen containers are uniquely designed to be leak proof and protect against biohazardous or hazardous material release to the environment
- Highly visible OSHA compliant labeling
- Sizes range from 7mL dermatology containers to 7.5 Liters operating room organ buckets
- Container openings range from 13mm to 300mm
- Filled with 10% neutral buffered formalin to 50% of container’s capacity
- Formalin waste designed to be treated in bulk using Neutralex or treated individually using Neutralexr tablets (patent pending).
- Nonsterile containers marked with a lot code and expiration date, 4 year shelf life
- Manufactured using ISO 13485 and FDA quality systems
Specifications
Specifications
| Product Type | Specimen Container Vial |
| Closure Included | Yes |
| Graduated | Yes |
| Material | Polypropylene |
| Sterility | Non-sterile |
| Capacity (English) | 3 oz., pre-filled 50% |
| Capacity (Metric) | 90 mL, pre-filled 50% |
| Closure Color | White |
| Closure Material | Polyethylene |
| Closure Size | 56 mm |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.