Learn More
CPC Scientific H-His-Ser-Asp-Ala-Leu-Phe-Thr-Asp-Thr-Tyr-Thr-Arg-Leu-Arg-Lys-Gln-Met-Ala-Met-Lys-Lys-Tyr-Leu-Asn-Ser-Val-Leu-Asn-NH2 (trifluoroacetate salt) 1MG
SDP
Supplier:  CPC Scientific VIP005B
Encompass
Vasoactive intestinal peptide derived from guinea pig. Though it differs from other mammalian species by four amino acids (positions 5, 9, 19, 26), this substitution does not have any impact on its biological activity.ONE-LETTER SEQUENCE: HSDALFTDTYTRLRKQMAMKKYLNSVLN-NH2MOLECULAR FORMULA: C147H239N43O42S2MOLECULAR WEIGHT:3344.91STORAGE CONDITIONS: -20 5CRESEARCH AREA: GastrointestinalREFERENCES: B.H. Du et al., BBRC, 128, 1093 (1985)
Catalog No. 50-211-0613
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.