Learn More
Bachem VIP (human, mouse, rat) Acetate
SDP
Supplier:  Bachem 4011611.0001
Encompass_Preferred
VIP (human, mouse, rat) Acetate, H-His-Ser-Asp-Ala-Val-Phe-Thr-Asp-Asn-Tyr-Thr-Arg-Leu-Arg-Lys-Gln-Met-Ala-Val-Lys-Lys-Tyr-Leu-Asn-Ser-Ile-Leu-Asn-NH2, 40077-57-4, C147H238N44O42S, Mr 3325.84, 1mg. Aviptadil, VIP, human, porcine, rat, ovine. VIP, a 28-amino acid peptide belonging to the secretin-glucagon-CRF family, is widely distributed in the central and peripheral nervous systems. Acting both as a neurotransmitter and as a hormone, VIP is involved in a wide variety of biological activities, including vaso- and bronchodilation, smooth muscle relaxation, and stimulation of secretion. An injectable formulation of this peptide in combination with the adrenergic drug phentolamine is expected to provide a new and effective alternative for erectile dysfunction patients.
Catalog No. NC9587718
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.