Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ VIPR1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PA3113
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PA3113 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PA3113 Supplier Invitrogen™ Supplier No. PA3113
Add to Cart
Edge
Add to Cart

Rabbit Polyclonal Antibody

PA3-113 detects VPAC1 Receptor in human samples. PA3-113 has been successfully used in Western blot, immunocytochemistry and immunohistochemistry (paraffin) procedures. By Western blot, this antibody detects an ~55-70 kDa protein representing VPAC1 Receptor in human samples. In immunohistochemistry, VPAC1 Receptor immunoreactivity is predominantly localized at the level of the plasma membrane. PA3-113 immunizing peptide corresponds to amino acid residues 438-457 from human VPAC1 Receptor.

The Vasoactive Intestinal Polypeptide Receptor 1 (VPAC1) is a member of a family of three G-protein coupled receptors which mediate VIP (vasoactive intestinal peptide) and PACAP (pituitary adenlyate cyclase activating peptide) function in a variety of tissues. Together, the VPAC1 and VPAC2 receptors compose a subtype of the VIP receptor. Expression of the VIP and PACAP receptors has been linked to the presence of human tumors.
TRUSTED_SUSTAINABILITY

Specifications

Antigen VIPR1
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration Conc. Not Determined
Conjugate Unconjugated
Formulation Whole serum with 0.05% sodium azide
Gene VIPR1
Gene Accession No. P32241, P97751
Gene Alias AV071699; HVR1; II; Pacap receptor; PACAP type II receptor; PACAP-R2; PACAP-R-2; pituitary adenylate cyclase activating polypeptide receptor, type II; pituitary adenylate cyclase-activating polypeptide type II receptor; Pvr2; RATVASREC; RDC1; type 1 vasoactive intestinal peptide receptor; V1RG; VAPC1; vasoactive intestinal peptide receptor 1; vasoactive intestinal polypeptide receptor 1; Vasopressive intestinal peptide receptor; VASREC; VIP and PACAP receptor 1; VIP receptor 1; VIP receptor subtype 1; VIP receptor, type I; VIP1 receptor; VIPR; Vipr1; VIP-R1; VIP-R-1; VIRG; VPAC1; VPAC1 receptor; VPAC1R; VPCAP1R
Gene Symbols VIPR1
Host Species Rabbit
Immunogen Synthetic peptide, KLH conjugated, corresponding to the residues T(438) R V S P G A R R S S S F Q A E V S L V(457) of human VPAC1 receptor.
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 22354, 7433
Target Species Human, Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.