Learn More
Cliawaived Inc ViraDx™ SARS-CoV-2/Flu A+B Rapid Antigen Test
SDP
Supplier:  Cliawaived Inc LUMCP0031
Encompass_Preferred
ViraDx™ SARS-CoV-2/Flu A+B Rapid Antigen Test is a lateral flow immunoassay intended for the in vitro rapid, simultaneous qualitative detection and differentiation of nucleocapsid protein antigen from SARS-CoV-2, influenza A and influenza B directly from anterior nasal or nasopharyngeal swab specimens collected from individuals, who are suspected of respiratory viral infection consistent with COVID-19 by their healthcare provider, within the first five (5) days of onset of symptoms, when tested at least twice over three days with at least 48 hours between tests. Clinical signs and symptoms of respiratory viral infection due to SARS-CoV-2 and influenza can be similar.
Catalog No. NC2610563
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.