Learn More
MALIX LLC The ViraDx™ SARS-CoV-2/Flu A+B Rapid Antigen Test detects and differentiates SARS-CoV-2, influenza A, and influenza B using nasal or nasopharyngeal swabs. Provides accurate results in 15 minutes. For in vitro diagnostic use, Rx only.
Supplier:  MALIX LLC VIRADXCOVFLU
Encompass
ViraDx™ POC Test for SARS-CoV-2, Flu A & B
The ViraDx™ SARS-CoV-2/Flu A+B Rapid Antigen Test is a lateral flow immunoassay designed for in vitro rapid, simultaneous qualitative detection and differentiation of nucleocapsid antigens from SARS-CoV-2, influenza A, and influenza B. It uses anterior nasal or nasopharyngeal swab specimens from patients suspected of COVID-19 by their healthcare provider within the first five days of symptom onset. This test provides efficient, rapid, and accurate results within 15 minutes, aiding in improved patient management decisions at the point-of-care. It is intended for in vitro diagnostic use and is for prescription use only (Rx).
Catalog No. NC3235646