Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Viral HIV-1 Gag p24 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. p-7113177
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB9539SP 25 μg
MAB9539100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. MAB9539SP Supplier R&D Systems Supplier No. MAB9539SP
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Monoclonal Antibody

HIV-1 Gag p24 Monoclonal specifically detects HIV-1 Gag p24 in Viral samples. It is validated for Western Blot.

Specifications

Antigen HIV-1 Gag p24
Applications Western Blot
Classification Monoclonal
Clone 2230C
Conjugate Unconjugated
Dilution Western Blot 0.1 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS.
Gene Accession No. Q77YG1
Host Species Rabbit
Immunogen E. coli-derived recombinant viral HIV-1 Gag p24 Pro149-Gly354 Accession # Q77YG1
Purification Method Protein A or G purified from cell culture supernatant
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 155030
Test Specificity Detects viral HIV-1 Gag p24 in Western blots.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Virus
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG
Show More Show Less

RUO

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.