Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Viral MIP-I Monoclonal Antibody (84420)
GREENER_CHOICE

Catalog No. MA561893
Encompass_Preferred
Change view
Click to view available options
Quantity:
500 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MA561893 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MA561893 Supplier Invitrogen™ Supplier No. MA561893
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Reconstitute at 0.5 mg/mL in sterile PBS.

MIP-1, also known as Macrophage Inflammatory Protein-1, is a chemokine encoded by the CCL3 gene. This protein is a member of the CC chemokine family and plays a critical role in the immune response by recruiting and activating various types of leukocytes, including macrophages, T cells, and dendritic cells, to sites of infection or injury. MIP-1 is involved in the inflammatory response, promoting the release of other cytokines and chemokines that amplify the immune reaction. It binds to specific receptors on the surface of target cells, such as CCR1 and CCR5, to exert its effects. Elevated levels of MIP-1 have been associated with various inflammatory and autoimmune conditions, including rheumatoid arthritis, multiple sclerosis, and inflammatory bowel disease. Additionally, MIP-1 is implicated in the pathogenesis of certain cancers and HIV infection, where it can influence tumor progression and viral replication. Understanding the regulation and function of MIP-1 is crucial for developing targeted therapies aimed at modulating immune responses and treating diseases characterized by excessive inflammation or immune dysregulation. Research into MIP-1 also provides insights into the broader mechanisms of chemokine-mediated cell signaling and its potential therapeutic applications.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Viral MIP-I
Applications Neutralization, Western Blot
Classification Monoclonal
Clone 84420
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with trehalose and no preservative
Host Species Mouse
Immunogen E. coli-derived recombinant human herpes virus-8 MIP-I, Ala25-Ala95.
Purification Method Protein A/G
Quantity 500 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Virus
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Lyophilized
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.