Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gibco™ Viral Production Medium, AGT™

Catalog No. A5147103
Change view
Click to view available options
Reconstituted Volume:
10 L
100 L
1 L
50 L
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Reconstituted Volume
A5147103 50 L
A5147101 1 L
A5147102 10 L
A5147104 100 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. A5147103 Supplier Gibco™ Supplier No. A5147103
Add to Cart
Edge
Add to Cart
Request a Quote

Gibco Viral Production Medium, AGT, is the dry-format version of CTS Viral Production Medium, a chemically defined, serum-free, protein-free, animal origin-free, phenol red-free medium.

  • Gibco Viral Production Medium, AGT is the dry-format version of CTS Viral Production Medium, a chemically defined, serum-free, protein-free, animal origin-free, phenol red-free medium
  • Medium is part of the CTS AAV-MAX Helper-Free AAV Production System for high-titer adeno-associated virus (AAV) production
  • The CTS AAV-MAX Helper-Free AAV Production System is a scalable, high-titer platform for suspension AAV production with research-grade and GMP versions to enable seamless transition from research to commercial manufacturing
  • Other components of the CTS AAV-MAX system include CTS Viral Production Cells 2.0, CTS AAV-MAX Transfection Kit, and CTS Viral-Plex Complexation Buffer.

Features of Viral Production Medium, AGT, include:

  • Supports growth and transfection of CTS Viral Production Cells 2.0 from shake flask to bioreactor scales
  • Helps enable production of high AAV titers, greater than 5 x 1010 vg/mL (unpurified)
  • Simplifies transfer to manufacturing scale due to Advanced Granulation Technology (AGT) enabling auto-pH and simplified dissolution
  • Formulated with GlutaMAX Supplement, no further supplementation required
  • Manufactured in conformity with cGMP for medical devices, 21 CFR, Part 820, and certified to ISO 13485. Drug Master File and Regulatory Support File available upon request.

Advanced Granulation Technology

  • Advanced Granulation Technology (AGT) helps simplify medium preparation at large scale with fast rehydration
  • A just-add water protocol, and low dust generation, creates less pressure build-up during sterile filtration
  • For more information on AGT media preparation at large scale, see Hydration and scale-up of AGT medium in HyPerforma and imPULSE Single-Use Mixers

Specifications

Cell Line Viral Production Cells 2.0
Cell Type HEK 293 cells
Concentration 25.6 g/L
Form Powder (AGT)
Product Type AAV Production Medium
Sterility Non-sterile
With Additives Glucose, GlutaMAX
Without Additives No Phenol Red, No Antibiotics
For Use With (Application) AAV Production
Product Line AAV-MAX
Quantity 50 L
pH 6.9 to 7.5
Reconstituted Volume 50 L
Content And Storage 50 L Ropack Bucket, store at 2°C to 8°C protected from light
Culture Type Suspension Cell Culture
Expression System Mammalian
Show More Show Less
What settings should I use to count my cells using the Vi-CELL XR Cell Counter?

You can refer to page 9 of the AAV-MAX Helper-Free AAV Production System User Guide (https://assets.thermofisher.com/TFS-Assets/LSG/manuals/MAN0019619_AAV-MAX_Helper-Free_AAV_ProductionSystem_UG.pdf) for detailed Vi-CELL XR settings. We recommend diluting cell count samples with PBS to eliminate precipitate artifacts that may be created by interaction with the Vi-CELL reagents.

Does Viral Production Medium, AGT contain a surfactant? Do I need to add extra surfactant for culturing CTS VPC2.0 in bioreactors ?

While the full formulation for Viral Production Medium, AGT is proprietary information that cannot be disclosed, we can confirm that the medium contains a surfactant and that no additional surfactant would need to be added to the medium to support your suspension CTS VPC2.0 in 2 L bench-top bioreactor to 50 L bioreactor.

What is the concentration of glucose in Viral Production Medium, AGT?

Viral Production Medium, AGT is formulated to contain glucose at a concentration of 6 g/L.

What type of filter is recommended to prepare liquid aseptic media from Viral Production Medium, AGT powder?

Viral Production Medium, AGT powder needs to be reconstituted in deionized distilled water or Water For Injection (WFI) for Cell Culture (Cat. No. A12873) and then immediately sterilized by membrane filtration (positive pressure recommended). A low protein binding filter (such as PVDF) with a pore size of 0.2 µM is recommended.

How does Viral Production Medium, AGT powder differ from the research-grade Viral Production Medium (Cat. No. A48179)?

GlutaMAX Supplement (Cat. No. 35050061) is already present in the one-part Viral Production Medium, AGT powder. Once rehydrated, the formulations of the two products are identical provided the research-grade Viral Production Medium (Cat No. A48179) has been supplemented with GlutaMAX Supplement at point of use.

How much Viral Production Medium, AGT powder do I weigh out for one liter of media?

25.6 grams of Viral Production Medium, AGT powder is required for 1 L of media. We do not advise preparing less than 1 L at a time to ensure consistent weighing and rehydration process.


Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.