Learn More
Cliawaived Inc KaiBiLi - Universal Transport System with 3mL Medium fill and a Nasopharyngeal Swab (400 swabs and vials per box)
SDP
Supplier:  Cliawaived Inc CLIAUTSKG
Encompass_Preferred
KaiBiLi - Universal Transport System with 3mL Medium and (1) Nasal Pharyngeal Swab. 102mm length vial contains 3ml of solution. Includes nasopharyngeal swab with synthetic fibers, plastic shaft, and break point at 80mm. The KaiBili UTS - Transport Medium is an FDA-listed collection and transport system suitable for collection, transport, maintenance and long term freeze storage of clinical specimens containing viruses, including Chlamydia and Mycoplasma.
Catalog No. NC2580201