Learn More
Pharmacal Research Laboratories Virkon-S broad-spectrum disinfectant/ cleaner
Supplier: Pharmacal Research Laboratories 030751F
Virkon-S is a broad-spectrum disinfectant/ cleaner for use in an animal research facility. It is fragrance-free and effective against numerous microorganisms affecting animals, including viruses, gram positive and gram-negative bacteria, fungi (molds and yeasts), and Mycoplasma. Virkon-S solutions are stable for 7 days after mixing. (50 tablets per Bottle)
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.